APO456797766 Apollo Scientific Ltd

4-Bromo-3-fluorothioanisole

  • MDL number MFCD09027283
  • Cas Number : 917562-25-5
  • Molecular Weight : 221.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Bromo-2-fluoro-4-(methylthio)benzene, 1-Bromo-2-fluoro-4-(methylsulphanyl)benzene, 4-Bromo-3-fluorophenyl methyl sulphide
Purity 96
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-3-fluorothioanisole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-3-fluorothioanisole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart