APO456797855 Apollo Scientific Ltd

4-Deoxy-4-fluoro-D-glucose

  • MDL number MFCD00077529
  • Cas Number : 29218-07-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189-190
Synonym 4-Fluoro-4-deoxy-d-glucopyranose
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Shipped with Ice Packs
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2915

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=2915
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart