APO456797883 Apollo Scientific Ltd

2-(Trifluoromethyl)phenyl isothiocyanate

  • MDL number MFCD00039644
  • Cas Number : 1743-86-8
  • Molecular Weight : 203.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Isothiocyanato-2-(trifluoromethyl)benzene
Purity 98
Density 1.346
Flash Point 105
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 95-96/10mm
Product Link https://store.apolloscientific.co.uk/product/2-trifluoromethylphenyl-isothiocyanate

Safety Information

signalword Warning
pcodes P260;P280;P312
MSDS https://store.apolloscientific.co.uk/product/2-trifluoromethylphenyl-isothiocyanate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart