APO456797976 Apollo Scientific Ltd

2-(Trifluoromethyl)benzylamine

  • MDL number MFCD00010297
  • Cas Number : 3048-01-9
  • Molecular Weight : 175.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(Aminomethyl)benzotrifluoride, [2-(Trifluoromethyl)phenyl]methylamine
Density 1.249
Flash Point 69
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 108-110/64mm
Product Link https://store.apolloscientific.co.uk/product/2-trifluoromethylbenzylamine

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-trifluoromethylbenzylamine
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart