APO456797979 Apollo Scientific Ltd

2-(Methylamino)-5-(trifluoromethyl)pyridine

  • MDL number MFCD08689790
  • Cas Number : 937602-15-8
  • Molecular Weight : 176.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 91-93
Iupac Name N-methyl-5-(trifluoromethyl)-2-pyridinamine
Inchi Code 1S/C7H7F3N2/c1-11-6-3-2-5(4-12-6)7(8,9)10/h2-4H,1H3,(H,11,12)
Inchi Key ALRNMRIFTCZDDH-UHFFFAOYSA-N

Description

Synonym N-Methyl-5-(trifluoromethyl)pyridin-2-amine, 6-(Methylamino)-alpha,alpha,alpha-trifluoro-3-picoline
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 91-93
Product Link https://store.apolloscientific.co.uk/product/2-methylamino-5-trifluoromethylpyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-methylamino-5-trifluoromethylpyridine
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart