APO456798052 Apollo Scientific Ltd

3,6-Difluorophthalic acid

  • MDL number MFCD03840499
  • Cas Number : 651-97-8
  • Molecular Weight : 202.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184 (dec.)
Purity 99
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/36-difluorophthalic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/36-difluorophthalic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart