APO456798064 Apollo Scientific Ltd

Tris(3-fluorophenyl)phosphine

  • MDL number MFCD00015722
  • Cas Number : 23039-94-3
  • Molecular Weight : 316.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 62-63
Iupac Name tris(3-fluorophenyl)phosphine
Inchi Code 1S/C18H12F3P/c19-13-4-1-7-16(10-13)22(17-8-2-5-14(20)11-17)18-9-3-6-15(21)12-18/h1-12H
Inchi Key CUTRINLXFPIWQB-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 62-63
Product Link https://store.apolloscientific.co.uk/product/tris3-fluorophenylphosphine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/tris3-fluorophenylphosphine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart