APO456798068 Apollo Scientific Ltd

1,2-Dichloro-3,3,3-trifluoroprop-1-ene

  • MDL number MFCD08461577
  • Cas Number : 431-27-6
  • Molecular Weight : 164.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -109.2
Synonym 1,2-Dichloro-3,3,3-trifluoropropene
Purity 98
Density 1.46
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 53-55
Product Link https://store.apolloscientific.co.uk/product/12-dichloro-333-trifluoroprop-1-ene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/12-dichloro-333-trifluoroprop-1-ene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart