APO456798124 Apollo Scientific Ltd

5-(Trifluoromethyl)-1H-indazole

  • MDL number MFCD08234904
  • Cas Number : 885271-64-7
  • Molecular Weight : 186.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 123-125
Iupac Name 5-(trifluoromethyl)-1H-indazole
Inchi Code 1S/C8H5F3N2/c9-8(10,11)6-1-2-7-5(3-6)4-12-13-7/h1-4H,(H,12,13)
Inchi Key WBHMPANPIGWXQV-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 123-125
Product Link https://store.apolloscientific.co.uk/product/5-trifluoromethyl-1h-indazole

Safety Information

signalword Warning
pcodes P271;P280;P261
MSDS https://store.apolloscientific.co.uk/product/5-trifluoromethyl-1h-indazole
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart