APO456798226 Apollo Scientific Ltd

2,6-Difluoro-4-hydroxybenzoic acid

  • MDL number MFCD03094500
  • Cas Number : 214917-68-7
  • Molecular Weight : 174.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170-171
Synonym 4-Carboxy-3,5-difluorophenol
Purity 97
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/26-difluoro-4-hydroxybenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/26-difluoro-4-hydroxybenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart