APO456798230 Apollo Scientific Ltd

2-Bromo-4-fluorothioanisole

  • MDL number MFCD09801042
  • Cas Number : 91524-70-8
  • Molecular Weight : 221.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-4-fluoro-1-(methylsulfanyl)benzene
Inchi Code 1S/C7H6BrFS/c1-10-7-3-2-5(9)4-6(7)8/h2-4H,1H3
Inchi Key XQGBAFLNAQHQNY-UHFFFAOYSA-N

Description

Synonym 2-Bromo-4-fluoro-1-(methylsulphanyl)benzene, 2-Bromo-4-fluorophenyl methyl sulphide
Purity 96%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-4-fluorothioanisole

Safety Information

signalword Warning
pcodes P280;P301+P312;P304+P340
MSDS https://store.apolloscientific.co.uk/product/2-bromo-4-fluorothioanisole
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart