APO456798307 Apollo Scientific Ltd

6-Methyl-4-(piperazin-1-yl)-2-(trifluoromethyl)quinoline

  • MDL number MFCD00728815
  • Cas Number : 544429-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-[6-Methyl-2-(trifluoromethyl)quinol-4-yl]piperazine
Purity 97
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/6-methyl-4-piperazin-1-yl-2-trifluoromethylquinoline-97

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-methyl-4-piperazin-1-yl-2-trifluoromethylquinoline-97
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart