APO456798387 Apollo Scientific Ltd

4-Fluoro-2-oxindole

  • MDL number MFCD04112528
  • Cas Number : 138343-94-9
  • Molecular Weight : 151.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-fluoro-1,3-dihydro-2H-indol-2-one
Inchi Code 1S/C8H6FNO/c9-6-2-1-3-7-5(6)4-8(11)10-7/h1-4,10-11H
Inchi Key SHIXDMKYVDHOFC-UHFFFAOYSA-N

Description

Synonym 1,3-Dihydro-4-fluoro-2H-indol-2-one, 4-Fluoroindolin-2-one
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-fluoro-2-oxindole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-fluoro-2-oxindole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart