APO456798436 Apollo Scientific Ltd

Perfluoro-3,6-dioxaoctane-1,8-dioic acid

  • MDL number MFCD03094537
  • Cas Number : 55621-21-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,2'-[(1,1,2,2-Tetrafluoro-1,2-ethanediyl)bis(oxy)]bis(difluoroacetic acid)
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 115/0.8mm
Product Link https://store.apolloscientific.co.uk/product/perfluoro-36-dioxaoctane-18-dioic-acid-98

Safety Information

signalword Danger
pcodes P260;P280;P310
MSDS https://store.apolloscientific.co.uk/product/perfluoro-36-dioxaoctane-18-dioic-acid-98
pictograms GHS05;GHS07
hstatements H314;H335
Availability details for
Quantity
  • Availability
  • Add to Cart