APO456798460 Apollo Scientific Ltd

2,3-Difluoro-4-hydroxybenzaldehyde

  • MDL number MFCD08235194
  • Cas Number : 676500-39-3
  • Molecular Weight : 158.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140-144
Purity 97
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/23-difluoro-4-hydroxybenzaldehyde

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/23-difluoro-4-hydroxybenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart