APO456798519 Apollo Scientific Ltd

1,1,2,3,3,3-Hexafluoropropyl dichloromethyl ether

  • MDL number MFCD03094547
  • Cas Number : 56860-82-3
  • Molecular Weight : 250.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 98
Product Link https://store.apolloscientific.co.uk/product/112333-hexafluoropropyl-dichloromethyl-ether

Safety Information

signalword Warning
pcodes P302+P352;P305+P351+P338;P362+P364
MSDS https://store.apolloscientific.co.uk/product/112333-hexafluoropropyl-dichloromethyl-ether
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart