APO456798635 Apollo Scientific Ltd

2-Bromo-2-fluoroacetamide

  • MDL number MFCD04038267
  • Cas Number : 430-91-1
  • Molecular Weight : 155.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 44
Synonym 2-Bromo-2-fluoroethanamide
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-2-fluoroacetamide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-2-fluoroacetamide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart