APO456798647 Apollo Scientific Ltd

Tetrafluorobenzene-1,2-diol

  • MDL number MFCD01075279
  • Cas Number : 1996-23-2
  • Molecular Weight : 182.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 68-69
Iupac Name 3,4,5,6-tetrafluoro-1,2-benzenediol
Inchi Code 1S/C6H2F4O2/c7-1-2(8)4(10)6(12)5(11)3(1)9/h11-12H
Inchi Key AQSZAISVBFUJQG-UHFFFAOYSA-N

Description

Synonym 1,2-Dihydroxy-3,4,5,6-tetrafluorobenzene, Tetrafluorocatechol, Perfluorobenzene-1,2-diol
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 68-69
Product Link https://store.apolloscientific.co.uk/product/tetrafluorobenzene-12-diol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/tetrafluorobenzene-12-diol
pictograms GHS07
hstatements H335
Availability details for
Quantity
  • Availability
  • Add to Cart