APO456876380 Apollo Scientific Ltd

2',4'-Dihydroxy-3'-methylpropiophenone

  • MDL number MFCD00010818
  • Cas Number : 63876-46-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124-130
Synonym 1-(2,4-Dihydroxy-3-methylphenyl)propan-1-one, 4-(Ethylcarbonyl)-2-methylbenzene-1,3-diol, 2,6-Dihydroxy-3-(ethylcarbonyl)toluene
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=17000

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=17000
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart