APO456876382 Apollo Scientific Ltd

2,4-Dichlorophenoxyacetic acid hydrazide

  • MDL number MFCD00051972
  • Cas Number : 28236-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150-152
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/24-dichlorophenoxyacetic-acid-hydrazide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/24-dichlorophenoxyacetic-acid-hydrazide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart