APO456876438 Apollo Scientific Ltd

5-Bromo-1,2,3,4-tetrahydroisoquinoline

  • MDL number MFCD08544279
  • Cas Number : 81237-69-6
  • Molecular Weight : 212.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-1234-tetrahydroisoquinoline

Safety Information

signalword Warning
pcodes P260;P280;P312
MSDS https://store.apolloscientific.co.uk/product/5-bromo-1234-tetrahydroisoquinoline
pictograms GHS07
hstatements H302+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart