APO456876445 Apollo Scientific Ltd

3,6-Dioxaoctanedioic acid

  • MDL number MFCD00054502
  • Cas Number : 23243-68-7
  • Molecular Weight : 178.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/36-dioxaoctanedioic-acid

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/36-dioxaoctanedioic-acid
pictograms GHS07
hstatements H302+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart