APO456876446 Apollo Scientific Ltd

N-Ethylglycine, N-BOC protected

  • MDL number MFCD00672508
  • Cas Number : 149794-10-5
  • Molecular Weight : 203.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 87-88
Synonym N-(tert-Butoxycarbonyl)-N-ethylglycine
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/n-ethylglycine-n-boc-protected

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/n-ethylglycine-n-boc-protected
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart