APO456876502 Apollo Scientific Ltd

1-[3-(Pyrrolidin-1-yl)prop-1-yl]homopiperazine

  • MDL number MFCD01320893
  • Cas Number : 199475-39-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-[3-(Pyrrolidin-1-yl)prop-1-yl]-1,4-diazepane, 1-[3-(1,4-Diazepan-1-yl)prop-1-yl]pyrrolidine
Purity 97
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=15705

Safety Information

signalword Danger
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=15705
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart