APO456876532 Apollo Scientific Ltd

7-Bromo-6-methyl-1H-indazole

  • MDL number MFCD17168273
  • Cas Number : 1257535-45-7
  • Molecular Weight : 211.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp >110
Iupac Name 7-bromo-6-methyl-1H-indazole
Inchi Code 1S/C8H7BrN2/c1-5-2-3-6-4-10-11-8(6)7(5)9/h2-4H,1H3,(H,10,11)
Inchi Key YQYJCEGYOVYERM-UHFFFAOYSA-N

Description

Melting Point >110
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/7-bromo-6-methyl-1h-indazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/7-bromo-6-methyl-1h-indazole
pictograms GHS07
hstatements H302;H312;H315;H319;H332;H335
Availability details for
Quantity
  • Availability
  • Add to Cart