APO456876546 Apollo Scientific Ltd

3-Amino-4-bromo-1H-pyrazole

  • MDL number MFCD00082728
  • Cas Number : 16461-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-140
Synonym 4-Bromo-1H-pyrazol-3-amine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-amino-4-bromo-1h-pyrazole

Safety Information

signalword Warning
pcodes P260;P280;P312
MSDS https://store.apolloscientific.co.uk/product/3-amino-4-bromo-1h-pyrazole
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart