APO456876709 Apollo Scientific Ltd

2-Bromo-3-fluorobenzotrifluoride

  • MDL number MFCD00061243
  • Cas Number : 104540-42-3
  • Molecular Weight : 243

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-1-fluoro-3-(trifluoromethyl)benzene
density 1.741
Inchi Code 1S/C7H3BrF4/c8-6-4(7(10,11)12)2-1-3-5(6)9/h1-3H
Inchi Key UERAGXKMOXUWPC-UHFFFAOYSA-N

Description

Purity 98%
Density 1.741
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-3-fluorobenzotrifluoride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-3-fluorobenzotrifluoride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart