APO456876734 Apollo Scientific Ltd

4'-(Difluoromethoxy)acetophenone

  • MDL number MFCD00042250
  • Cas Number : 83882-67-1
  • Molecular Weight : 186.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 18-19
Purity 98
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 70-72/0.5mm
Product Link https://store.apolloscientific.co.uk/product/4-difluoromethoxyacetophenone

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-difluoromethoxyacetophenone
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart