APO456876771 Apollo Scientific Ltd

3-[(Trifluoromethyl)thio]phenylacetic acid

  • MDL number MFCD00236350
  • Cas Number : 239080-04-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53-55
Synonym 3-[(Trifluoromethyl)sulphanyl]phenylacetic acid, 3-(Carboxymethyl)phenyl trifluoromethyl sulphide
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-trifluoromethylthiophenylacetic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-trifluoromethylthiophenylacetic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart