APO456876780 Apollo Scientific Ltd

2-Chloro-4-(trifluoromethyl)benzoic acid

  • MDL number MFCD06208178
  • Cas Number : 23228-45-7
  • Molecular Weight : 224.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 114-117
Iupac Name 2-chloro-4-(trifluoromethyl)benzoic acid
Inchi Code 1S/C8H4ClF3O2/c9-6-3-4(8(10,11)12)1-2-5(6)7(13)14/h1-3H,(H,13,14)
Inchi Key DTIJZEUKFYGSEC-UHFFFAOYSA-N

Description

Synonym 4-Carboxy-3-chlorobenzotrifluoride
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 114-117
Product Link https://store.apolloscientific.co.uk/product/2-chloro-4-trifluoromethylbenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-4-trifluoromethylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart