APO456876781 Apollo Scientific Ltd

4-(Difluoromethyl)benzyl alcohol

  • MDL number MFCD20488855
  • Cas Number : 444915-77-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym [4-(Difluoromethyl)phenyl]methanol, 4-(Hydroxymethyl)benzal fluoride
Physical Form Solid-Low Melt
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=59905

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=59905
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart