APO456876804 Apollo Scientific Ltd

3,4-Difluoro-5-methylbenzoic acid

  • MDL number MFCD09832332
  • Cas Number : 1017778-60-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4,5-Difluoro-m-toluic acid, 5-Carboxy-2,3-difluorotoluene
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/34-difluoro-5-methylbenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/34-difluoro-5-methylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart