APO456876806 Apollo Scientific Ltd

(Trifluoromethoxy)acetic acid

  • MDL number MFCD11858185
  • Cas Number : 69105-00-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(Trifluoromethoxy)ethanoic acid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=60686

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=60686
Availability details for
Quantity
  • Availability
  • Add to Cart