APO456876822 Apollo Scientific Ltd

5-Bromo-2-chloro-4-(trifluoromethyl)pyridine

  • MDL number MFCD11100520
  • Cas Number : 823221-93-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Bromo-2-chloro-alpha,alpha,alpha-trifluoro-4-picoline
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 86-88/19mm
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-chloro-4-trifluoromethylpyridine

Safety Information

signalword Warning
pcodes P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-chloro-4-trifluoromethylpyridine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart