APO456876826 Apollo Scientific Ltd

2-Fluoro-4-(trifluoromethoxy)aniline

  • MDL number MFCD15527448
  • Cas Number : 123572-58-7
  • Molecular Weight : 195.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-fluoro-4-(trifluoromethoxy)aniline
Inchi Code 1S/C7H5F4NO/c8-5-3-4(1-2-6(5)12)13-7(9,10)11/h1-3H,12H2
Inchi Key HDDJDTDRGKHRBG-UHFFFAOYSA-N

Description

Purity 99%
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-4-trifluoromethoxyaniline

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-4-trifluoromethoxyaniline
pictograms GHS06
hstatements H302;H311+H331;H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart