APO456876902 Apollo Scientific Ltd

3-Fluoro-5-hydroxybenzoic acid

  • MDL number MFCD11846515
  • Cas Number : 860296-12-4
  • Molecular Weight : 156.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 214-215
Synonym 3-Carboxy-5-fluorophenol
Purity 98
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-5-hydroxybenzoic-acid

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-5-hydroxybenzoic-acid
pictograms GHS05;GHS07
hstatements H302;H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart