APO456876925 Apollo Scientific Ltd

2',4',6'-Trifluoropropiophenone

  • MDL number MFCD00061224
  • Cas Number : 220141-69-5
  • Molecular Weight : 188.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/246-trifluoropropiophenone

Safety Information

signalword Warning
pcodes P302+P352;P304+P340;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/246-trifluoropropiophenone
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart