APO456876940 Apollo Scientific Ltd

2-Bromo-3-fluorophenol

  • MDL number MFCD04037220
  • Cas Number : 443-81-2
  • Molecular Weight : 191

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-3-fluorophenol
Inchi Code 1S/C6H4BrFO/c7-6-4(8)2-1-3-5(6)9/h1-3,9H
Inchi Key LMFRSLRJXLATRL-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-3-fluorophenol

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-3-fluorophenol
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart