APO456876943 Apollo Scientific Ltd

3-Fluorothiophene-2-carboxylic acid

  • MDL number MFCD03426911
  • Cas Number : 32431-84-8
  • Molecular Weight : 146.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-fluoro-2-thiophenecarboxylic acid
Inchi Code 1S/C5H3FO2S/c6-3-1-2-9-4(3)5(7)8/h1-2H,(H,7,8)
Inchi Key WPHRBUAOSDHRDS-UHFFFAOYSA-N

Description

Synonym 2-Carboxy-3-fluorothiophene
Storage temp. Store at 2-8
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fluorothiophene-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluorothiophene-2-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart