APO456988984 Apollo Scientific Ltd

3-tert-Butoxycarbonylamino-2-methyl-propionic acid

  • MDL number MFCD02094137
  • Cas Number : 16948-10-0
  • Molecular Weight : 203.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 88-90
Iupac Name N-(tert-butoxycarbonyl)-2-methyl-beta-alanine
Inchi Code 1S/C9H17NO4/c1-6(7(11)12)5-10-8(13)14-9(2,3)4/h6H,5H2,1-4H3,(H,10,13)(H,11,12)
Inchi Key GDQRNRYMFXDGMS-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 88-90
Product Link https://store.apolloscientific.co.uk/product/3-tert-butoxycarbonylamino-2-methyl-propionic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-tert-butoxycarbonylamino-2-methyl-propionic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart