APO456989046 Apollo Scientific Ltd

3�Bromo-2�4�difluoroacetophenone

  • MDL number MFCD13689251
  • Cas Number : 1210824-63-7
  • Molecular Weight : 235.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3-bromo-2,4-difluorophenyl)ethanone
Inchi Code 1S/C8H5BrF2O/c1-4(12)5-2-3-6(10)7(9)8(5)11/h2-3H,1H3
Inchi Key NOQZJPYTXYUMAU-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-24-difluoroacetophenone

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-24-difluoroacetophenone
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart