APO456989111 Apollo Scientific Ltd

Methyl 5-bromo-2-formylbenzoate

  • MDL number MFCD16036575
  • Cas Number : 1016163-89-5
  • Molecular Weight : 243.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 5-bromo-2-formylbenzoate
Inchi Code 1S/C9H7BrO3/c1-13-9(12)8-4-7(10)3-2-6(8)5-11/h2-5H,1H3
Inchi Key ZYGJVCDDZZSSEE-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-5-bromo-2-formylbenzoate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-5-bromo-2-formylbenzoate
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart