APO456989185 Apollo Scientific Ltd

Imidazo[1,2-a]pyridine-8-carbonitrile

  • Cas Number : 136117-70-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. TBC
Shipping Temperature TBC
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=74207

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=74207
Availability details for
Quantity
  • Availability
  • Add to Cart