APO458378485 Apollo Scientific Ltd

4-Cyclopropyl-benzenemethanol

  • MDL number MFCD06802675
  • Cas Number : 454678-87-6
  • Molecular Weight : 148.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-cyclopropyl-benzenemethanol

Safety Information

signalword Warning
pcodes P280;P302+P352
MSDS https://store.apolloscientific.co.uk/product/4-cyclopropyl-benzenemethanol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart