APO458378555 Apollo Scientific Ltd

3-Amino-1-benzyl-pyrrolidine-3-carboxylic acid

  • MDL number MFCD06410530
  • Cas Number : 145602-87-5
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-amino-1-benzyl-3-pyrrolidinecarboxylic acid
Inchi Code 1S/C12H16N2O2/c13-12(11(15)16)6-7-14(9-12)8-10-4-2-1-3-5-10/h1-5H,6-9,13H2,(H,15,16)
Inchi Key IXBOFOMZFVXPTF-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-amino-1-benzyl-pyrrolidine-3-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-1-benzyl-pyrrolidine-3-carboxylic-acid
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart