APO458378598 Apollo Scientific Ltd

1-Boc-4-ethynyl-1H-pyrazole

  • MDL number MFCD16293741
  • Cas Number : 1093193-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Methyl-2-propanyl 4-ethynyl-1H-pyrazole-1-carboxylate
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=74481

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=74481
pictograms GHS07
hstatements H302;H312;H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart