APO458378624 Apollo Scientific Ltd

1-(4-Fluorophenyl)cyclopropylmethanol

  • MDL number MFCD07374455
  • Cas Number : 198976-40-8
  • Molecular Weight : 166.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-4-fluorophenylcyclopropylmethanol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1-4-fluorophenylcyclopropylmethanol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart