APO458378670 Apollo Scientific Ltd

3�Bromo-5�fluoroacetophenone

  • MDL number MFCD11847746
  • Cas Number : 105515-20-6
  • Molecular Weight : 217.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3-bromo-5-fluorophenyl)ethanone
Inchi Code 1S/C8H6BrFO/c1-5(11)6-2-7(9)4-8(10)3-6/h2-4H,1H3
Inchi Key MZDXPUDHZBCYGS-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-5-fluoroacetophenone

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-5-fluoroacetophenone
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart