APO458378682 Apollo Scientific Ltd

2-Bromo-4-iodonitrobenzene

  • MDL number MFCD18391757
  • Cas Number : 860603-83-4
  • Molecular Weight : 327.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-4-iodo-1-nitrobenzene
Inchi Code 1S/C6H3BrINO2/c7-5-3-4(8)1-2-6(5)9(10)11/h1-3H
Inchi Key GPVOKEUHKNSWFV-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-4-iodonitrobenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-4-iodonitrobenzene
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart