APO458378683 Apollo Scientific Ltd

3-Bromo-2-iodonitrobenzene

  • MDL number MFCD16660176
  • Cas Number : 32337-96-5
  • Molecular Weight : 327.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-bromo-2-iodo-3-nitrobenzene
Inchi Code 1S/C6H3BrINO2/c7-4-2-1-3-5(6(4)8)9(10)11/h1-3H
Inchi Key BJOZUBWLKNXFFK-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-2-iodonitrobenzene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-2-iodonitrobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart